missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYC Tag Mouse, CoraLite 647, Clone: 1A5A2, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech CL647-60003-100UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The MYC tag antibody from Proteintech is a mouse monoclonal antibody generated with peptide of MYC tag. This antibody recognizes recombinant protein antigen. The MYC tag antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite 647.
Spécification
MYC Tag | |
1A5A2 | |
CoraLite 647 | |
Mouse | |
Protein G Purification | |
Primary | |
Liquid |
Flow Cytometry (Intracellular Staining), Immunofluorescence | |
1000 μg/mL | |
PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3 | |
Peptide | |
100 μL | |
Store at -20°C | |
IgG1 |