Learn More
MTUS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 13436-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The antibody can recognize all isoforms except isoform 5 (85-90 kDa). The antibody tested HomoDimer isoforms (80 kDa/110-120/280 kDa) in Jurkat cell.
The protein belongs to the FKBP-type peptidyl-prolyl cis/trans isomerase family. It is located in endoplasmic reticulum and acts as molecular chaperones.Spécification
MTUS1 | |
Polyclonal | |
Unconjugated | |
MTUS1 | |
AT2 receptor binding protein, ATBP, ATIP, GK1, KIAA1288, MP44, MTSG1, MTUS1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
102103, 306487, 57509 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5HZI1, Q6IMY1, Q9ULD2 | |
MTUS1 | |
MTUS1 Fusion Protein Ag4242 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |