Learn More
MRPL24 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 16224-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein which is more than twice the size of its E. coli counterpart. Sequence analysis identified two transcript variants that encode the same protein.Spécification
MRPL24 | |
Polyclonal | |
Unconjugated | |
MRPL24 | |
L24mt, MRP L18, MRP L24, MRPL24 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
295224, 67707, 79590 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.18 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q66H47, Q96A35, Q9CQ06 | |
mrpl24 | |
MRPL24 Fusion Protein Ag9277 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |