missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPL14 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 15040-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein. A pseudogene corresponding to this gene is found at 17p13.3.Spécification
MRPL14 | |
Polyclonal | |
Unconjugated | |
MRPL14 | |
L14mt, L32mt, MRP L14, MRP L32, MRPL14, MRPL32, RMPL32, RPML32 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
301250, 64928, 68463 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.24 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q6P1L8, Q7M0E7, Q9D1I6 | |
MRPL14 | |
MRPL14 Fusion Protein Ag6808 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |