Learn More
MP Biomedicals™ Casein sodium salt
Casein sodium salt White to off-white powder
114.48€
Spécification
Forme | Powder |
---|---|
Couleur | White to off-white |
Synonyme | Sodium Caseinate |
Description
- Casein protein has numerous experimental applications including use as a blocking agent in immunochemistry, recovery of enzyme activity from SDS extracted samples, and as a substrate for protease and kinase assays.
- Casein sodium salt from bovine milk is a phosphoprotein. There are four main types of Casein which make up approximately 80% of the total protein in bovine milk: α-s1 Casein, α-s2 Casein, β-Casein, and κ-Casein.
- Casein is proposed to be the main protective constituent in milk.
Spécification
Powder | |
White to off-white | |
Sodium Caseinate |