Learn More
MOV10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 10370-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Probable RNA helicase. Required for RNA-mediated gene silencing by the RNA-induced silencing complex (RISC). Required for both miRNA-mediated translational repression and miRNA- mediated cleavage of complementary mRNAs by RISC. Also required for RNA-directed transcription and replication of the human hepatitis delta virus (HDV). Interacts with small capped HDV RNAs derived from genomic hairpin structures that mark the initiation sites of RNA-dependent HDV RNA transcription.Spécification
MOV10 | |
Polyclonal | |
Unconjugated | |
MOV10 | |
fSAP113, gb110, KIAA1631, MOV10, Putative helicase MOV 10 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
17454, 310756, 4343 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.55 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P23249, Q9HCE1 | |
Mov10 | |
MOV10 Fusion Protein Ag0384 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |