missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MANBAL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 16028-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
MANBAL is a protein coding gene.Spécification
MANBAL | |
Polyclonal | |
Unconjugated | |
MANBAL | |
MANBAL, Protein MANBAL | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
499934, 63905, 69161 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.28 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9D8X0, Q9NQG1 | |
MANBAL | |
MANBAL Fusion Protein Ag8888 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |