missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAGEB18 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 17494-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The melanoma-associated antigen (MAGE) family consists of a number of antigens recognized by cytotoxic T lymphocytes. The MAGE genes were initially isolated from different kinds of tumors and, based on their virtually exclusive tumor-specific expression in adult tissues, they have been used as targets for cancer immunotherapy. MAGE genes encode for tumor-rejection antigens that are expressed in tumors of different histologic types and in normal testis and placenta. MAGE-B18 (melanoma-associated antigen B18) is a 343 amino acid protein that contains one MAGE domain and may be involved in tumorigenesis. The gene encoding MAGE-B18 maps to human chromosome X, which contains nearly 153 million base pairs and houses over 1,000 genes.Spécification
MAGEB18 | |
Polyclonal | |
Unconjugated | |
MAGEB18 | |
MAGE B18 antigen, MAGEB18, melanoma antigen family B, 18 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
286514 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q96M61 | |
MAGEB18 | |
MAGEB18 Fusion Protein Ag11624 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |