missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIMS1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 55336-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Pinch, also designated particularly interesting new Cys-His protein or NY-REN-48, is a focal adhesion protein that is a component of the ILK-PINCH complex. This complex is a major part of the growth factor and integrin signaling pathway. Pinch is involved in cell differentiation, proliferation and survival by acting as an effector of integrin and growth factor signaling. It is a cytoplasmic protein expressed in most tissues and consists of five LIM domains, a nuclear localization signal and a nuclear export signal. The PINCH-1/ILK complex is regulated by a PINCH-1 related protein PINCH-2, which also forms a complex with ILK.Spécification
LIMS1 | |
Polyclonal | |
Unconjugated | |
LIMS1 | |
LIMS1, PINCH, PINCH 1, PINCH1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
3987 | |
-20°C | |
Liquid |
Western Blot | |
0.63 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P48059 | |
LIMS1 | |
Peptide | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |