Learn More
Labconco™ Armoire de stockage standard Protector™
L’armoire de stockage standard Labconco Protector™ peut être utilisée pour le stockage de fournitures et de petits instruments de laboratoire.
Marque:  Labconco™ 9900200
Code produit. 17935012
  • 1935.00€ / 1 pièce
Découvrez plus d'offres spéciales
Includes: Four leveling feet and one 8 in. filler panel to increase cabinet depth from 22 to 30 in. .
Description
Description
The Labconco Protector™ Standard Storage Cabinet may be used for storage of supplies and small laboratory instruments.
  • Support loads up to 800 lb. (363kg)
  • Durable epoxy-coated steel construction
  • Attractive glacier white exterior complements laboratory casework and Labconco fume hoods
  • Manual-closing, non-locking door(s)
Informations sur la commande
Work surface and interior shelf kits are sold separately
Spécification
Spécification
Storage Cabinet
Manual Close
35.5 to 36.75 in.
Four Leveling Feet and One 8 in. D Filler Panel to Increase Cabinet Depth from 22 to 30 in.
800 lb.
2
155 lb.
30 in.
Epoxy-coated Steel
Blanc
Ideal for storage and venting of small laboratory instrument and mild acid and other reagent
90.17 to 93.34 cm
22 in.
362.87 kg
Epoxy
70.3 kg
76.2 cm