Learn More
KIF12 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 12035-1-AP-20UL
 Afficher plus de versions de ce produit
Code produit. 16819513
  • 145.00€ / 20µL
Découvrez plus d'offres spéciales
Description
Description
KIF12 encodes a member of the kinesin superfamily of microtubule-associated molecular motors with functions related to the microtubule cytosekelton. Members of this superfamily play important roles in intracellular transport and cell division. A similar protein in mouse functions in the beta cell antioxidant signaling cascade, acting as a scaffold for the transcription factor specificity protein 1 (Sp1). Mice that lack this gene exhibit beta cell oxidative stress resulting in hypoinsulinemic glucose intolerance.
Spécification
Spécification
KIF12
Polyclonal
Unconjugated
KIF12
KIF12, kinesin family member 12, Kinesin like protein KIF12, RP11 56P10.3
Rabbit
Antigen Affinity Chromatography
RUO
113220, 16552, 313254
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q96FN5, Q9D2Z8
Kif12
KIF12 Fusion Protein Ag2707
20 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Suggestions de produits
Suggestions de produits
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Sonderangebote
Sonderangebote