missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIF12 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 12035-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
KIF12 encodes a member of the kinesin superfamily of microtubule-associated molecular motors with functions related to the microtubule cytosekelton. Members of this superfamily play important roles in intracellular transport and cell division. A similar protein in mouse functions in the beta cell antioxidant signaling cascade, acting as a scaffold for the transcription factor specificity protein 1 (Sp1). Mice that lack this gene exhibit beta cell oxidative stress resulting in hypoinsulinemic glucose intolerance.Spécification
KIF12 | |
Polyclonal | |
Unconjugated | |
KIF12 | |
KIF12, kinesin family member 12, Kinesin like protein KIF12, RP11 56P10.3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
113220, 16552, 313254 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q96FN5, Q9D2Z8 | |
Kif12 | |
KIF12 Fusion Protein Ag2707 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |