Learn More
KEAP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 10503-2-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
This antibody is a rabbit polyclonal antibody raised against residues near the C terminus of human KEAP1.
This gene encodes a protein containing KELCH-1 like domains, as well as a BTB/POZ domain. Kelch-like ECH-associated protein 1 interacts with NF-E2-related factor 2 in a redox-sensitive manner and the dissociation of the proteins in the cytoplasm is followed by transportation of NF-E2-related factor 2 to the nucleus. This interaction results in the expression of the catalytic subunit of gamma-glutamylcysteine synthetase. Two alternatively spliced transcript variants encoding the same isoform have been found for this gene.Spécification
KEAP1 | |
Polyclonal | |
Unconjugated | |
KEAP1 | |
Cytosolic inhibitor of Nrf2, INrf2, KEAP1, Kelch like protein 19, KIAA0132, KLHL19 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
117519, 50868, 9817 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P57790, Q14145, Q9Z2X8 | |
Keap1 | |
KEAP1 Fusion Protein Ag0779 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |