missing translation for 'onlineSavingsMsg'
Learn More
Learn More
INT11 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 15860-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Chromatin assembly factor I (CAF-I) is required for the assembly of histone octamers onto newly-replicated DNA. CAF-I is composed of three protein subunits, p50, p60, and p150. The protein encoded by this gene corresponds to the p60 subunit and is required for chromatin assembly after replication. The encoded protein is differentially phosphorylated in a cell cycle-dependent manner. In addition, it is normally found in the nucleus except during mitosis, when it is released into the cytoplasm. This protein is a member of the WD-repeat HIR1 family and may also be involved in DNA repair.Spécification
INT11 | |
Polyclonal | |
Unconjugated | |
INTS11 | |
CPSF3 like protein, CPSF3L, CPSF73L, INT11, Integrator complex subunit 11, INTS11, RC 68, RC68 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
54973, 71957 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5TA45, Q9CWS4 | |
INTS11 | |
INT11 Fusion Protein Ag8365 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |