Learn More
Proteintech™ HumanKine™ Recombinant Human IL-17 (IL-17A) Protein
HumanKine™ cytokines and growth factors are human cell expressed proteins in HEK293 cells
Marque:  Proteintech™ HZ-1113-1000UG
Code produit. 16881316
  • 2087.00€ / 1000µg
Découvrez plus d'offres spéciales
Description
Description
  • Results in authentic human proteins with native glycosylation, folding and increased stability, activity and purity
Spécification
Spécification
3605
1000 μg
Briefly centrifuge the vial before opening. It is recommended to reconstitute the protein in sterile 1 x PBS containing 0.1% endotoxin-free recombinant human serum albumin (HSA).
Typically ≤ 2 ng/mL EC50
Lyophilized
30 to 35 kDa, homodimer
Human
Lyophilized proteins are stable for 1 year from the date of receipt if stored between -20 to -80°C. Upon reconstitution, it is recommended the solution be stored at 4°C for short term or at -20 to -80°C for long term. Repeated freeze thaw cycles should be avoided with reconstituted products.
Human embryonic kidney 293 (HEK293) human cell expression system
> 95%