missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HTF9C Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 16199-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The protein encoded by TRMT2A is of unknown function. However, it is orthologous to the mouse Trmt2a gene and contains an RNA methyltransferase domain. Expression of TRMT2A varies during the cell cycle, with aberrant expression being a possible biomarker in certain breast cancers. Several transcript variants encoding two different isoforms have been found for TRMT2A.Spécification
HTF9C | |
Polyclonal | |
Unconjugated | |
TRMT2A | |
HTF9C, TRMT2A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
15547, 27037, 287953 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8BNV1, Q8IZ69 | |
Trmt2a | |
HTF9C Fusion Protein Ag9112 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |