missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSPB8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 15287-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The protein encoded by this gene belongs to the superfamily of small heat-shock proteins containing a conservative alpha-crystallin domain at the C-terminal part of the molecule. The expression of this gene in induced by estrogen in estrogen receptor-positive breast cancer cells, and this protein also functions as a chaperone in association with Bag3, a stimulator of macroautophagy. Thus, this gene appears to be involved in regulation of cell proliferation, apoptosis, and carcinogenesis, and mutations in this gene have been associated with different neuromuscular diseases, including Charcot-Marie-Tooth disease.Spécification
HSPB8 | |
Polyclonal | |
Unconjugated | |
HSPB8 | |
Alpha crystallin C chain, CMT2L, CRYAC, DHMN2, E2 induced gene 1 protein, E2IG1, H11, heat shock 22kDa protein 8, Heat shock protein beta 8, HMN2, HMN2A, HSP22, HSPB8, Protein kinase H11 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
113906, 26353, 80888 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9EPX0, Q9JK92, Q9UJY1 | |
Hspb8 | |
HSPB8 Fusion Protein Ag7456 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |