missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSD17B8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16752-1-AP-20UL
This item is not returnable.
View return policy
Description
NAD-dependent 17-beta-hydroxysteroid dehydrogenase with highest activity towards estradiol. Has very low activity towards testosterone. The heteroteramer with CBR4 has NADH-dependent 3- ketoacyl-acyl carrier protein reductase activity. May play a role in biosynthesis of fatty acids in mitochondria.Specifications
HSD17B8 | |
Polyclonal | |
Unconjugated | |
HSD17B8 | |
17 beta HSD 8, D6S2245E, FABG, FABGL, H2 KE6, HKE6, HSD17B8, Ke 6, KE6, Protein Ke6, RING2, SDR30C1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14979, 361802, 7923 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P50171, Q6MGB5, Q92506 | |
Hsd17b8 | |
HSD17B8 Fusion Protein Ag10226 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |