missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HN1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 14914-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
HN1 (hematopoietic- and neurologic-expressed sequence 1) is a highly conserved protein that is expressed in developing and regenerating tissues. Overexpression of HN1 has been reported in various tumors like glioma and breast cancer. Hn1 has also been reported as a marker for human ovarian carcinoma and can distinguish epithelial ovarian carcinoma cells from normal ovarian surface epithelial cells. This collective information suggests that HN1 is involved in processes associated with cell proliferation, repair and/or growth.Spécification
HN1 | |
Polyclonal | |
Unconjugated | |
JPT1 | |
Androgen regulated protein 2, ARM2, HN1, HN1A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
15374, 287828, 51155 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.43 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P97825, Q6AXU6, Q9UK76 | |
Jpt1 | |
HN1 Fusion Protein Ag6712 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |