Learn More
HLA-DQA2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 55072-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The antibody is specific to HLA-DQA2.
HLA-DQA2 belongs to the HLA class II alpha chain family. The encoded protein forms a heterodimer with a class II beta chain. It is located in intracellular vesicles and plays a central role in the peptide loading of MHC class II molecules by helping to release the CLIP molecule from the peptide binding site. Class II molecules are expressed in antigen presenting cells (B lymphocytes, dendritic cells, macrophages) and are used to present antigenic peptides on the cell surface to be recognized by CD4 T-cells.Spécification
HLA-DQA2 | |
Polyclonal | |
Unconjugated | |
HLA-DQA2 | |
DX alpha chain, HLA DQA1, HLA DQA2, HLA DXA, HLA-DQA2, MHC class II DQA2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
3118 | |
-20°C | |
Liquid |
Western Blot | |
0.51 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P01906 | |
HLA-DQA2 | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |