missing translation for 'onlineSavingsMsg'
Learn More

HDAC6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 16167-1-AP-20UL

 Afficher plus de versions de ce produit

Code produit. 16815704

  • 145.00€ / 20µL

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Découvrez plus d'offres spéciales
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Description

Description

This antibody is specific to HDAC6. It does not cross react with other HDACs.

Responsible for the deacetylation of lysine residues on the N-terminal part of the core histones (H2A, H2B, H3 and H4). Histone deacetylation gives a tag for epigenetic repression and plays an important role in transcriptional regulation, cell cycle progression and developmental events. Histone deacetylases act via the formation of large multiprotein complexes. Plays a central role in microtubule-dependent cell motility via deacetylation of tubulin.
Spécification

Spécification

HDAC6
Polyclonal
Unconjugated
HDAC6
HD6, HDAC6, histone deacetylase 6, JM21, KIAA0901
Rabbit
Antigen Affinity Chromatography
RUO
10013, 15185, 84581
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.39 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9UBN7, Q9Z2V5
Hdac6
Peptide
20 μL
Primary
Mouse, Human, Rat
Antibody
IgG
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions