missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Cat Whole Serum (Forensic) Polyclonal Antibody, MP Biomedicals
Marque: MP Biomedicals 0855999
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Host Species: Goat
Species Reactivity: Cat
Spécification
Whole Serum | |
Unconjugated | |
2 mL | |
Feline |
Polyclonal | |
Goat | |
Primary | |
Lyophilized |