Learn More
FUT3 Mouse anti-Human, Unconjugated, Clone: 2A6G7, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 67344-1-IG-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The FUT3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human FUT3. This antibody recognizes human antigen. The FUT3 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Spécification
FUT3 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
2A6G7 | |
Unconjugated | |
20 μL | |
2525 | |
IgG1 |