Learn More
FLNA Mouse anti-Human, Unconjugated, Clone: 1G4H3, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 67133-1-IG-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The FLNA antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human FLNA. This antibody recognizes human antigen. The FLNA antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Spécification
FLNA | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
1G4H3 | |
Unconjugated | |
150 μL | |
2316 | |
IgG2a |