missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ENDOG Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 22148-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The protein encoded by this gene is a nuclear encoded endonuclease that is localized in the mitochondrion. The encoded protein is widely distributed among animals and cleaves DNA at GC tracts. This protein is capable of generating the RNA primers required by DNA polymerase gamma to initiate replication of mitochondrial DNA.Spécification
ENDOG | |
Polyclonal | |
Unconjugated | |
ENDOG | |
Endo G, ENDOG, endonuclease G, Endonuclease G, mitochondrial | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
13804, 2021, 362100 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O08600, Q14249 | |
Endog | |
ENDOG Fusion Protein Ag17739 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |