Learn More
Cole-Parmer™ Heating Block for Electrothermal™ RS-250D-S Shaking Reaction Stations
Click to view available options
Capacité:
4 x 96-well PTFE Microplates
40 x 24 mm Tubes
96 x 16 mm Tubes
Description
- Compatible with Electrothermal RS9000 Reaction Stations
Spécification
Spécification
Description | Heating Block for Reaction System |
À utiliser avec (équipement) | Tubes |
Capacité | 40 x 24 mm Tubes |
No. of Places | 40 trous |
Type | Heating Block |
Garantie | 1 Year |