Learn More
DOK2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 14087-1-AP-150UL
 Afficher plus de versions de ce produit
Code produit. 16822574
  • 410.00€ / 150µL
Découvrez plus d'offres spéciales
Description
Description
ERK2 (also known as MAPK1) is a member of the MAP kinase subfamily of Ser/Thr protein kinases. MAP kinases, also known as extracellular signal-regulated kinases (ERKs), act as an integration point for multiple biochemical signals, and are involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation and development. The activation of this kinase requires its phosphorylation by upstream kinases. Upon activation, this kinase translocates to the nucleus of the stimulated cells, where it phosphorylates nuclear targets.
Spécification
Spécification
DOK2
Polyclonal
Unconjugated
DOK2
Docking protein 2, docking protein 2, 56kDa, DOK2, p56(dok 2), p56DOK, p56dok 2
Rabbit
Antigen Affinity Chromatography
RUO
13449, 290361, 9046
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.41 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O60496, O70469
Dok2
DOK2 Fusion Protein Ag5218
150 μL
Primary
Human, Mouse, Rat