missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DLEC1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00€ - 399.00€
Spécification
Antigène | DLEC1 |
---|---|
Concentration | 0.27 mg/mL |
Applications | Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Immunoprecipitation, Immunocytochemistry |
Classification | Polyclonal |
Conjugué | Unconjugated |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
16819514
|
Proteintech
20027-1-AP-20UL |
20 μL |
140.00€
20µL |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
16809514
|
Proteintech
20027-1-AP-150UL |
150 μL |
399.00€
150µL |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
Description
This gene contains 37 exons, spans approximately 59-kb, and is located in the 3p22-p21.3 chromosomal segment that is commonly deleted in various carcinomas. Several alternatively spliced transcripts have been observed that contain disrupted coding regions and likely encode nonfunctional proteins. Aberrant transcription of this gene may be involved in carcinogenesis of the lung, esophagus, and kidney.Spécification
DLEC1 | |
Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Immunoprecipitation, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q8BLA1, Q9Y238 | |
320256, 9940 | |
Peptide | |
Primary | |
-20°C | |
DLEC1 |
0.27 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
DLC 1, DLC1, DLEC1, F56 | |
Dlec1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |