Learn More
DCTPP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 16684-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The protein encoded by this gene is dCTP pyrophosphatase, which converts dCTP to dCMP and inorganic pyrophosphate. The encoded protein also displays weak activity against dTTP and dATP, but none against dGTP. This protein may be responsible for eliminating excess dCTP after DNA synthesis and may prevent overmethylation of CpG islands. Three transcript variants, one protein-coding and the other two non-protein coding, have been found for this gene.Spécification
DCTPP1 | |
Polyclonal | |
Unconjugated | |
DCTPP1 | |
CDA03, dCTP pyrophosphatase 1, dCTPase 1, DCTPP1, Deoxycytidine triphosphatase 1, RS21C6, XTP3TPA | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
192252, 66422, 79077 | |
-20°C | |
Liquid |
Immunocytochemistry, Western Blot, Immunofluorescence | |
0.38 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q91VC0, Q9H773, Q9QY93 | |
Dctpp1 | |
DCTPP1 Fusion Protein Ag10015 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |