missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTBP1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 10972-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
This gene encodes a member of the arginine/serine-rich family of splicing factors. The encoded protein functions in mRNA processing. Alternatively spliced transcript variants have been described.Spécification
CTBP1 | |
Polyclonal | |
Unconjugated | |
CTBP1 | |
BARS, C terminal binding protein 1, CTBP, CTBP1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
13016, 1487 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Western Blot, Immunoprecipitation | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O88712, Q13363 | |
Ctbp1 | |
CTBP1 Fusion Protein Ag1425 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |