Learn More
CLYBL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 17314-1-AP-20UL
Code produit. 16847184
  • 140.00€ / 20µL
Découvrez plus d'offres spéciales
Description
Description
CLYBL, also named as CLB, is a Mitochondrial citramalyl-CoA lyase indirectly involved in the vitamin B12 metabolism. It is a human mitochondrial enzyme of unknown function that is found in multiple eukaryotic taxa and conserved to bacteria. It acts as a beta-methylmalate synthase in vitro, by mediating conversion of glyoxylate and propionyl-CoA to beta-methylmalate. There are two isoforms with MW 31-33 kDa and 35-37 kDa.
Spécification
Spécification
CLYBL
Polyclonal
Unconjugated
CLYBL
citrate lyase beta like, CLB, CLYBL
Rabbit
Antigen Affinity Chromatography
RUO
171425, 306198, 69634
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q5I0K3, Q8N0X4, Q8R4N0
CLYBL
CLYBL Fusion Protein Ag11283
20 μL
Primary
Mouse, Rat, Human