missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CISD1 Rabbit anti-Human, Mouse, Rat, Zebrafish, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Spécification
Antigène | CISD1 |
---|---|
Concentration | 0.23 mg/mL |
Applications | Immunohistochemistry (Paraffin), Immunofluorescence, Flow Cytometry, Immunocytochemistry, Immunoprecipitation |
Classification | Polyclonal |
Conjugué | Unconjugated |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
16855444
|
Proteintech
16006-1-AP-20UL |
20 μL |
145.00€
20µL |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
16845444
|
Proteintech
16006-1-AP-150UL |
150 μL |
410.00€
150µL |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
Description
This antibody recognizing MitoNEET (calculated 12 kDa) as a 17 kDa protein may be due to its posttranslational modification or metal binding activity.
This gene encodes a protein with a CDGSH iron-sulfur domain and has been shown to bind a redox-active [2Fe-2S] cluster. The encoded protein has been localized to the outer membrane of mitochondria and is thought to play a role in regulation of oxidation. Genes encoding similar proteins are located on chromosomes 4 and 17, and a pseudogene of this gene is located on chromosome 2.Spécification
CISD1 | |
Immunohistochemistry (Paraffin), Immunofluorescence, Flow Cytometry, Immunocytochemistry, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Mouse, Zebrafish, Human, Rat | |
B0K020, Q91WS0, Q9NZ45 | |
294362, 393577, 52637, 55847 | |
CISD1 Fusion Protein Ag8680 | |
Primary | |
-20°C | |
CISD1 |
0.23 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
C10orf70, CDGSH iron sulfur domain 1, CISD1, MDS029, mitoNEET, Mnt, ZCD1 | |
Cisd1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |