Learn More
CD62L (L-Selectin) Rat anti-Mouse, CoraLite 647, Clone: MEL-14, Proteintech
Rat Monoclonal Antibody
Marque:  Proteintech CL647-65123-100TESTS
Code produit. 16861136
  • 120.51€ / 100 tests
Découvrez plus d'offres spéciales
Description
Description
The CD62L (L-Selectin) antibody from Proteintech is a rat monoclonal antibody generated with c3h/eb mouse b lymphoma 38c-13 of mouse CD62L (L-Selectin). This antibody recognizes mouse antigen. The CD62L (L-Selectin) antibody has been validated for the following applications: IF, FC analysis. This antibody is coralite™647.
Spécification
Spécification
CD62L (L-Selectin)
Monoclonal
5 μL/Test
Rat
Primary
Mouse
Immunofluorescence, Flow Cytometry
MEL-14
CoraLite 647
100 Tests
20343
IgG2a