Learn More
CD62L (L-Selectin) Rat anti-Mouse, CoraLite 647, Clone: MEL-14, Proteintech
Rat Monoclonal Antibody
Marque: Proteintech CL647-65123-100TESTS
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The CD62L (L-Selectin) antibody from Proteintech is a rat monoclonal antibody generated with c3h/eb mouse b lymphoma 38c-13 of mouse CD62L (L-Selectin). This antibody recognizes mouse antigen. The CD62L (L-Selectin) antibody has been validated for the following applications: IF, FC analysis. This antibody is coralite™647.
Spécification
CD62L (L-Selectin) | |
Monoclonal | |
5 μL/Test | |
Rat | |
Primary | |
Mouse |
Immunofluorescence, Flow Cytometry | |
MEL-14 | |
CoraLite 647 | |
100 Tests | |
20343 | |
IgG2a |