Learn More
CD44 Rat anti-Human, PE, Clone: IM7, Proteintech
Rat Monoclonal Antibody
Marque:  Proteintech PE-65117-100UG
Code produit. 16811716
  • 45.00€ / 100µg
Découvrez plus d'offres spéciales
Description
Description
The CD44 antibody from Proteintech is a rat monoclonal antibody generated with dexamethasone-induced myeloid leukemia m1 cells of mouse CD44. This antibody recognizes human, mouse antigen. The CD44 antibody has been validated for the following applications: FC analysis. This antibody is pe.
Spécification
Spécification
CD44
Monoclonal
0.2 mg/mL
Rat
Primary
Human, Mouse
Flow Cytometry
IM7
PE
100 μg
12505
IgG2b