Learn More
CD44 Rat anti-Human, PE, Clone: IM7, Proteintech
Rat Monoclonal Antibody
Marque: Proteintech PE-65117-100UG
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The CD44 antibody from Proteintech is a rat monoclonal antibody generated with dexamethasone-induced myeloid leukemia m1 cells of mouse CD44. This antibody recognizes human, mouse antigen. The CD44 antibody has been validated for the following applications: FC analysis. This antibody is pe.
Spécification
CD44 | |
Monoclonal | |
0.2 mg/mL | |
Rat | |
Primary | |
Human, Mouse |
Flow Cytometry | |
IM7 | |
PE | |
100 μg | |
12505 | |
IgG2b |