Learn More
CD4 Rat anti-Mouse, PE, Clone: RM4-5, Proteintech
Rat Monoclonal Antibody
Marque:  Proteintech PE-65141-100UG
Code produit. 16891726
  • 37.00€ / 100µg
Découvrez plus d'offres spéciales
Description
Description
The CD4 antibody from Proteintech is a rat monoclonal antibody generated with balb/c mouse thymocytes of mouse CD4. This antibody recognizes mouse antigen. The CD4 antibody has been validated for the following applications: FC analysis. This antibody is pe.
Spécification
Spécification
CD4
Monoclonal
0.2 mg/mL
Rat
Primary
Mouse
Flow Cytometry
RM4-5
PE
100 μg
12504
IgG2a