Learn More
CD38 Rat anti-Mouse, CoraLite 647, Clone: 90, Proteintech
Rat Monoclonal Antibody
Marque: Proteintech CL647-65059-100TESTS
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The CD38 antibody from Proteintech is a rat monoclonal antibody generated with mouse bone marrow pre-b cells of mouse CD38. This antibody recognizes mouse antigen. The CD38 antibody has been validated for the following applications: IF, FC analysis. This antibody is coralite™647.
Spécification
CD38 | |
Monoclonal | |
5 μL/Test | |
Rat | |
Primary | |
Mouse |
Immunofluorescence, Flow Cytometry | |
90 | |
CoraLite 647 | |
100 Tests | |
12494 | |
IgG2a |