Learn More
CD38 Rat anti-Mouse, CoraLite 647, Clone: 90, Proteintech
Rat Monoclonal Antibody
Marque:  Proteintech CL647-65059-100TESTS
Code produit. 16861106
  • 124.00€ / 100 tests
Découvrez plus d'offres spéciales
Description
Description
The CD38 antibody from Proteintech is a rat monoclonal antibody generated with mouse bone marrow pre-b cells of mouse CD38. This antibody recognizes mouse antigen. The CD38 antibody has been validated for the following applications: IF, FC analysis. This antibody is coralite™647.
Spécification
Spécification
CD38
Monoclonal
5 μL/Test
Rat
Primary
Mouse
Immunofluorescence, Flow Cytometry
90
CoraLite 647
100 Tests
12494
IgG2a