Learn More
CD27 Mouse anti-Human, CoraLite 647, Clone: O323, Proteintech
Mouse Monoclonal Antibody
Marque:  Proteintech CL647-65132-100TESTS
Code produit. 16891136
  • 136.00€ / 100 tests
Découvrez plus d'offres spéciales
Description
Description
The CD27 antibody from Proteintech is a mouse monoclonal antibody generated with n/a of human CD27. This antibody recognizes human antigen. The CD27 antibody has been validated for the following applications: FC analysis. This antibody is coralite™647.
Spécification
Spécification
CD27
Monoclonal
5 μL/Test
Mouse
Primary
Human
Flow Cytometry
O323
CoraLite 647
100 Tests
939
IgG1