Learn More
CD20 Mouse anti-Human, CoraLite 647, Clone: 4A7G3, Proteintech
Mouse Monoclonal Antibody
Marque:  Proteintech CL647-60271-100UL
Code produit. 16891096
  • 367.00€ / 100µL
Découvrez plus d'offres spéciales
Description
Description
The CD20 antibody from Proteintech is a mouse monoclonal antibody generated with fusion protein of human CD20. This antibody recognizes human antigen. The CD20 antibody has been validated for the following applications: FC, IF analysis. This antibody is coralite™647.
Spécification
Spécification
CD20
Monoclonal
CoraLite 647
100 μL
931
IgG2b
Immunofluorescence, Flow Cytometry
4A7G3