Learn More
CD20 Mouse anti-Human, CoraLite 647, Clone: 4A7G3, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech CL647-60271-100UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The CD20 antibody from Proteintech is a mouse monoclonal antibody generated with fusion protein of human CD20. This antibody recognizes human antigen. The CD20 antibody has been validated for the following applications: FC, IF analysis. This antibody is coralite™647.
Spécification
CD20 | |
Monoclonal | |
CoraLite 647 | |
100 μL | |
931 | |
IgG2b |
Immunofluorescence, Flow Cytometry | |
4A7G3 | |
Mouse | |
Primary | |
Human |