Learn More
CD11b Mouse anti-Human, FITC, Clone: ICRF44, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech FITC-65116-100TESTS
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The CD11b antibody from Proteintech is a mouse monoclonal antibody generated with n/a of human CD11b. This antibody recognizes human antigen. The CD11b antibody has been validated for the following applications: FC analysis. This antibody is fitc.
Spécification
CD11b | |
Monoclonal | |
5 μL (1 μg)/Test | |
Mouse | |
Primary | |
Human |
Flow Cytometry | |
ICRF44 | |
FITC | |
100 Tests | |
3684 | |
IgG1 |