Learn More
Inhibiteur de caspase 9 III, Thermo Scientific Chemicals
Code produit. 15446939
Click to view available options
Quantité:
1 mg
5 mg
A potent and irreversible inhibitor of caspase-9
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Identifiants chimiques
CAS 403848-57-7
Formule moléculaire C24H35ClN6O9
Synonyme Ac-LEHD-CMK ;
Spécification
Quantité 5 mg
Description Blanc
Contenu et stockage -30°C à -10°C
À utiliser avec (application) Un inhibiteur puissant et irréversible de la caspase-9
Poids de la formule 587.02
Forme Poudre