missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Carbonate, ≥99.0%, ACS Reagent, Honeywell Fluka™
Click to view available options
Quantité:
100 g
500 g
1 kg
2.5 kg
Conditionnement:
Poly Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Spécification
Spécification
Nom chimique ou matériau | Calcium Carbonate(Caco3) |
Point de fusion | 825°C |
Couleur | White |
Forme physique | Powder |
Quantité | 500 g |
Formule linéaire | CaCO3 |
Numéro UN | NONH for all modes of transport |
Beilstein | 8008338 |
Nom IUPAC | calcium;carbonate |
Poids de la formule | 100.09g/mol |
Afficher plus |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'