Learn More
Solution tampon pH 11, Thermo Scientific Chemicals Specpure™
29.75€ - 82.41€
Spécification
Nom chimique ou matériau Buffer solution
CAS 1336-21-6
Couleur Incolore
Forme physique Liquide
Formule moléculaire H5NO
Consulter d'autres spécifications
Produits 3
Code produit Marque Quantité Prix Quantité et disponibilité  
Code produit Marque Quantité Prix Quantité et disponibilité  
11306099
Afficher les documents Détails de la promotion
Thermo Scientific Alfa Aesar
042424.AP
500 mL
29.75€
500mL
11394588
Afficher les documents Détails de la promotion
Thermo Scientific Alfa Aesar
042424.K2
1 L
46.63€
1L
11384588
Afficher les documents Détails de la promotion
Thermo Scientific Alfa Aesar
042424.K7
4 L
82.41€
4L
Description
Description
Buffer solution (pH 11) is used as a means of keeping pH at a nearly constant value in different chemical applications. Buffer solution is used in alcoholic fermentation, leather tanning, electroplating, organic synthesis, sugar and paper manufacturing.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécification
Spécification
Buffer solution
Incolore
H5NO
MFCD00066650
VHUUQVKOLVNVRT-UHFFFAOYSA-N
35.05
CHEBI:18219
1336-21-6
Liquide
Odorless
Miscible with water.
N.O
14923
Vidéos
Sécurité et traitement
Sécurité et traitement
missing translation for 'tsca' : Oui
Recommended Storage : Température ambiante