Learn More
BL21(DE3)-lysate Rabbit anti-Bacteria, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 10558-1-IG-20UL
 Afficher plus de versions de ce produit
Code produit. 16897013
  • 145.00€ / 20µL
Découvrez plus d'offres spéciales
Description
Description
Escherichia coli B serves as a research model for studying phage sensitivity, restriction-modification systems, and bacterial evolution, and also as a workhorse for protein expression in life science laboratories and in the biotech industry. Characteristics such as protease deficiency, low acetate production at a high level of glucose, and enhanced permeability (probably due to a simple cell surface) make E. coli B a desirable host for the production of genetically engineered proteins. BL21(DE3) has been very widely used to express recombit proteins. Comparison of BL21(DE3) and another B strain REL606 shows they differ in length by 72. 304 bp and have 426 single nucleotide polymorphisms, all of which have been accounted for by various laboratory manipulations. Differences between B strains and K12 include the absence of flagellar component genes, the DNA cytosine methylase dcm, and ompT in BL21(DE3). B strains may have an additional type II secretion system not found in K12. Other differences include differential distribution of insertions sequences and other changes resulting from horizontal gene transfer. BL21(DE3) also carries a DE3 recombit phage harboring the T7 RNA polymerase gene that can direct high-level expression of cloned genes under the control of the T7 promoter.
Spécification
Spécification
BL21(DE3)-lysate
Polyclonal
Unconjugated
BL21(DE3) lysate
BL21(DE3)-lysate Fusion Protein Ag0048
20 μL
Primary
-20°C
Liquid
Western Blot
0.7 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Rabbit
Protein A
RUO
Bacteria