Learn More
ARL13B Mouse anti-Human, CoraLite 594, Clone: 1H6C3, Proteintech
Mouse Monoclonal Antibody
Marque:  Proteintech CL594-66739-100UL
Code produit. 16891066
  • 367.00€ / 100µL
Découvrez plus d'offres spéciales
Description
Description
The ARL13B antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ARL13B. This antibody recognizes human, canine antigen. The ARL13B antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Spécification
Spécification
ARL13B
Monoclonal
2000 μg/mL
Mouse
Primary
Human, Canine
Immunofluorescence
1H6C3
CoraLite 594
100 μL
200894
IgG2a