missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APLP1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 12305-2-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
This gene encodes a member of the highly conserved amyloid precursor protein gene family. The encoded protein is a membrane-associated glycoprotein that is cleaved by secretases in a manner similar to amyloid beta A4 precursor protein cleavage. This cleavage liberates an intracellular cytoplasmic fragment that may act as a transcriptional activator. The encoded protein may also play a role in synaptic maturation during cortical development. Alternatively spliced transcript variants encoding different isoforms have been described.Spécification
APLP1 | |
Polyclonal | |
Unconjugated | |
APLP1 | |
Amyloid like protein 1, APLP, APLP 1, APLP1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11803, 333 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P51693, Q03157 | |
APLP1 | |
APLP1 Fusion Protein Ag2951 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |