Learn More
RAB11A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marque: Invitrogen 715300
454.10 EUR valable jusqu'au 2025-05-31
Utilisez le code promo "25977" pour bénéficier de cette offre.
Description
This polyclonal antibody reacts specifically with the human Rab11A protein. Cross-reactivity with other Rab family members has not been detected. Positive control: Human HEK-293 cells
RAB11A Polyclonal antibody specifically detects RAB11A in Canine, Human, Mouse, Non-human Primate, Rabbit, Rat samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunomicroscopy, Immunoprecipitation, Western Blot.
Spécification
RAB11A | |
Polyclonal | |
Unconjugated | |
RAB11A | |
24KG; RAB 11A, member oncogene family; RAB11; rab-11; rab11 GTP-binding protein; Rab11a; RAB11a, member RAS oncogene family; ras-related protein Rab-11A; tubulovesicle-associated protein; YL8 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100009072, 403957, 53869, 81830, 8766 | |
-20°C | |
Liquid | |
HM 505 Cryostat |
ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunomicroscopy, Immunoprecipitation, Western Blot | |
0.25 mg/mL | |
PBS with 0.1% sodium azide; pH 7.4 | |
P62490, P62491, P62492, P62493, P62494 | |
RAB11A | |
Synthetic peptide derived from the C-terminal hypervariable region of the human Rab11a sequence. This peptide shares 10 of 17 residues with the related Rab11b protein. | |
100 μg | |
Primary | |
Canine, Human, Mouse, Non-human Primate, Rabbit, Rat | |
Antibody | |
IgG |
Certificats
Un numéro de lot est requis pour afficher les résultats sur les certificats. Pour retrouver le numéro de lot sur des commandes passées, utiliser le menu suivi de commande.
Numéro de lot | Type de certificat | Date | Catalog Number |
---|---|---|---|
AC409990 | Certificat d’analyse | 04/04/2025 |