missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Substrat d’oxyde d’aluminium, 10x10x0,5 mm, deux côtés polis, plan C., Thermo Scientific™
53.10€ - 630.00€
Identifiants chimiques
CAS | 1344-28-1 |
---|---|
Formule moléculaire | Al2O3 |
Poids moléculaire (g/mol) | 101.96 |
Numéro MDL | MFCD00003424 |
Clé InChI | PNEYBMLMFCGWSK-UHFFFAOYSA-N |
Synonyme | aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum |
CID PubChem | 9989226 |
Nom IUPAC | trioxidandiide de dialuminium(3+) |
SMILES | [O--].[O--].[O--].[Al+3].[Al+3] |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Identifiants chimiques
1344-28-1 | |
101.96 | |
PNEYBMLMFCGWSK-UHFFFAOYSA-N | |
9989226 | |
[O--].[O--].[O--].[Al+3].[Al+3] |
Al2O3 | |
MFCD00003424 | |
aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum | |
trioxidandiide de dialuminium(3+) |
Spécification
Aluminum oxide substrate | |
1344-28-1 | |
MFCD00003424 | |
PNEYBMLMFCGWSK-UHFFFAOYSA-N | |
trioxidandiide de dialuminium(3+) | |
9989226 |
10x10x0.5mm, polished two sides, C plane | |
Al2O3 | |
aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum | |
[O--].[O--].[O--].[Al+3].[Al+3] | |
101.96 |
Sécurité et traitement
missing translation for 'einecsNumber' : 215-691-6
missing translation for 'rtecsNumber' : BD1200000
missing translation for 'tsca' : Oui
Recommended Storage : Température ambiante