Learn More
Tube Al-23 pour serpentins chauffants, dia. ext. : 27 mm, dia. int. de : 20 mm, inclinaison : 7,5, Thermo Scientific™
148.00€ - 673.00€
Spécification
Nom chimique ou matériau | Al-23 Tube for Heating Coils |
---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
11313148
|
Thermo Scientific Alfa Aesar
033205.DM |
100 mm | |||||||
11333148
|
Thermo Scientific Alfa Aesar
033205.DR |
200 mm | |||||||
11303148
|
Thermo Scientific Alfa Aesar
033205.DW |
300 mm | |||||||
11393138
|
Thermo Scientific Alfa Aesar
033205.ED |
500 mm | |||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Spécification
Al-23 Tube for Heating Coils |
Sécurité et traitement
missing translation for 'tsca' : U
Recommended Storage : Ambient temperatures