Learn More
Tige ronde solide Al-23, Dia. : 1mm, Thermo Scientific™
Marque: Thermo Scientific Alfa Aesar 032923.DW
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Spécification
Al-23 Solid Round Rod | |
300 mm | |
Tige, ronde, diam. : 1 mm |