missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetyl CoA synthetase Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Spécification
Antigène | Acetyl CoA synthetase |
---|---|
Concentration | 0.3 mg/mL |
Applications | Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugué | Unconjugated |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
16855574
|
Proteintech
16087-1-AP-20UL |
20 μL |
145.00€
20µL |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
16845574
|
Proteintech
16087-1-AP-150UL |
150 μL |
410.00€
150µL |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
Description
This gene encodes a protein with a sterol sensing domain (SSD) and seven WD domains. In the presence of cholesterol, this protein binds to sterol regulatory element binding proteins (SREBPs) and mediates their transport from the ER to the Golgi. The SREBPs are then proteolytically cleaved and regulate sterol biosynthesis.Spécification
Acetyl CoA synthetase | |
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Rat, Human, Mouse | |
Q9NR19, Q9QXG4 | |
311569, 55902, 60525 | |
Acetyl CoA synthetase Fusion Protein Ag9121 | |
Primary | |
-20°C | |
ACSS2 |
0.3 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
ACAS2, AceCS, Acetate CoA ligase, Acetyl CoA synthetase, ACS, ACSA, ACSS2, Acyl activating enzyme | |
Acss2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |